missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM130/GOLGA2 Antibody (JE42-53), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32394
This item is not returnable.
View return policy
Description
GM130/GOLGA2 Monoclonal antibody specifically detects GM130/GOLGA2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| GM130/GOLGA2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 | |
| 130 kDa cis-Golgi matrix protein, GM130 autoantigen, GM130Golgi matrix protein GM130, golgi autoantigen, golgin subfamily a, 2, golgin A2, Golgin subfamily A member 2, golgin-95, MGC20672, SY11 protein | |
| Recombinant protein within human GM130/GOLGA2 aa 1-100 (N terminal). (Uniprot: Q08379) | |
| 100 μL | |
| Cellular Markers, Golgi Apparatus Markers | |
| 2801 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| JE42-53 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction