missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM-CSF Antibody Pair (HRP), Novus Biologicals™
Solid phase sandwich ELISA for the quantitative determination of Human GM-CSF.
£292.00 - £605.00
Specifications
| Antigen | GM-CSF |
|---|---|
| Dilution | Sandwich ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Host Species | Mouse |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18606161
|
Novus Biologicals
NBP2-78844-5Plates |
5 Plates |
£292.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18211617
|
Novus Biologicals
NBP2-78844-15PLATES |
15 Plates |
£605.00
Pack of 15 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GM-CSF Monoclonal specifically detects GM-CSF in Human samples. It is validated for ELISA, Sandwich ELISA.Specifications
| GM-CSF | |
| Monoclonal | |
| Mouse | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim | |
| CSF2 | |
| This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human GM-CSF. |
| Sandwich ELISA | |
| HRP | |
| Biologically Active Proteins, Cytokine Research, Immunology | |
| 1437 | |
| Storage of components varies. See protocol for specific instructions. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title