missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GM-CSF Antibody (otilimab), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28866-1mg
This item is not returnable.
View return policy
Description
GM-CSF Monoclonal antibody specifically detects GM-CSF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| GM-CSF | |
| Monoclonal | |
| Unconjugated | |
| P04141 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| otilimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim | |
| CSF2/ GM-CSF | |
| 1 mg | |
| Biologically Active Proteins, Cytokine Research, Immunology | |
| 1437 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction