missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GlyT1/SLC6A9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-03237-100UL
This item is not returnable.
View return policy
Description
GlyT1/SLC6A9 Polyclonal specifically detects GlyT1/SLC6A9 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GlyT1/SLC6A9 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6536 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| DKFZp547A1118, GlyT1, GlyT-1, GLYT1glyT-1, sodium- and chloride-dependent glycine transporter 1, solute carrier family 6 (neurotransmitter transporter, glycine), member 9, Solute carrier family 6 member 9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human GlyT1/SLC6A9 (NP_964012.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction