Learn More
GlyRS-IN-1, MedChemExpress
MedChemExpress GlyRS-IN-1 is a glycyl-tRNA synthase (GlyRS) inhibitor extracted from patent WO 2017066459 A1. GlyRS-IN-1 can also inhibit the growth of bacteria.
£600.16 - £960.74
Specifications
Chemical Name or Material | GlyRS-IN-1 |
---|---|
CAS | 112921-11-6 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C12H17N7O7S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16466967
![]() |
Medchem Express
HY-108940 |
5 mg |
£600.16
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16444248
![]() |
Medchem Express
HY-108940 |
10 mg |
£960.74
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GlyRS-IN-1 is a glycyl-tRNA synthase (GlyRS) inhibitor extracted from patent WO 2017066459 A1. GlyRS-IN-1 can also inhibit the growth of bacteria.Specifications
GlyRS-IN-1 | |
Off-White | |
C12H17N7O7S | |
O[C@@H]1[C@@H](COS(=O)(NC(CN)=O)=O)O[C@@H](N2C(N=CN=C3N)=C3N=C2)[C@@H]1O | |
403.37 | |
Research | |
Cancer-programmed cell death |
112921-11-6 | |
Research | |
DMSO : 100 mg/mL (247.91 mM; Need ultrasonic) | |
403.37 | |
98.14% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : -20°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
Recommended Storage : -20°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.