missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLYR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550669
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total GLYR1 protein.
GLYR1 may have oxidoreductase activity. It regulates p38 MAP kinase activity by mediating stress activation of p38alpha/MAPK14 and specifically regulating MAPK14 signaling. It indirectly promotes phosphorylation of MAPK14 and activation of ATF2. The phosphorylation of MAPK14 requires upstream activity of MAP2K4 and MAP2K6. It is recruited on chromatin, recognizes and binds trimethylated 'Lys-36' of histone H3 (H3K36me3).
Specifications
| GLYR1 | |
| Polyclonal | |
| Unconjugated | |
| GLYR1 | |
| 2810419J22Rik; 3930401k13rik; 3-hydroxyisobutyrate dehydrogenase-like protein; AW545332; BM045; Cytokine-like nuclear factor N-PAC; glyoxylate reductase 1 homolog; glyoxylate reductase 1 homolog (Arabidopsis); GLYR1; HIBDL; hNDF; NDF; NP60; Npac; N-pac; nuclear protein 60 kDa; nuclear protein 60kDa; nuclear protein NP60; Nuclear protein of 60 kDa; Nucleosome-destabilizing factor; Putative oxidoreductase GLYR1; RGD1309459 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 360477, 74022, 84656 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1.4 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q49A26, Q5RKH0, Q922P9 | |
| GLYR1 | |
| Synthetic peptide corresponding to a region derived from internal residues of human glyoxylate reductase 1 homolog (Arabidopsis). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction