missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Glypican 6 Recombinant Rabbit Monoclonal Antibody (8H34L5)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ 703424

Product Code. 17156288

  • £404.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is predicted to react with Mouse, Rat, Bovine.

The glypicans comprise a family of glycosylphosphatidylinositol-anchored heparan sulfate proteoglycans. The glypicans have been implicated in the control of cell growth and division. Glypican 6 is a putative cell surface coreceptor for growth factors, extracellular matrix proteins, proteases and anti-proteases.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Glypican 6
Recombinant Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9Y625
Gpc6
Peptides corresponding to human GPC6 [1) aa358-aa372 2) aa78-aa86].
100 μg
Primary
Human
Antibody
IgG
Western Blot, Immunocytochemistry
8H34L5
Unconjugated
Gpc6
glypican 6; glypican proteoglycan 6; glypican-6; GPC6; OMIMD1; Secreted glypican-6; UNQ369/PRO705
Rabbit
Protein A
RUO
10082
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.