missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glypican 5 Antibody (297716) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2607JF525
This item is not returnable.
View return policy
Description
Glypican 5 Monoclonal antibody specifically detects Glypican 5 in Human,Mouse samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| Glypican 5 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| bA93M14.1, glypican 5, glypican proteoglycan 5, glypican-5 | |
| Mouse myeloma cell line NS0-derived recombinant human Glypican 5, Glu25-Thr554, Accession # P78333 | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Western Blot, CyTOF | |
| 297716 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2262 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction