missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glypican 4 Antibody (51E3), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-49892PECY7
This item is not returnable.
View return policy
Description
Glypican 4 Monoclonal specifically detects Glypican 4 in Human samples. It is validated for Flow Cytometry.
Specifications
| Glypican 4 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| GPC4 | |
| Recombinant human GPC4(401-529aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Anti-human GPC4 mAb, clone AT51E3, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human GPC4 protein. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| 51000 | |
| Flow Cytometry | |
| glypican 4, glypican proteoglycan 4, glypican-4, K-glypicandJ900E8.1 (glypican 4) | |
| Mouse | |
| Protein A purified | |
| Cancer, Stem Cells | |
| 2239.0 | |
| Human | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction