missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glypican 4 Antibody (51E3), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-49892B
This item is not returnable.
View return policy
Description
Glypican 4 Monoclonal specifically detects Glypican 4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Glypican 4 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| GPC4 | |
| Recombinant human GPC4(401-529aa) purified from E. coli | |
| 100 μL | |
| Cancer, Stem Cells | |
| 2239 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 51000 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| glypican 4, glypican proteoglycan 4, glypican-4, K-glypicandJ900E8.1 (glypican 4) | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Anti-human GPC4 mAb, clone AT51E3, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human GPC4 protein. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction