missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Glypican 3 Recombinant Protein Antigen

Product Code. 18432322 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432322 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432322 Supplier Novus Biologicals™ Supplier No. NBP185226PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 2719
Molecular Weight (g/mol) 29 kDa
Quantity 0.1 mL
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias DGSX, glypican 3, glypican proteoglycan 3, glypican-3, GTR2-2, heparan sulphate proteoglycan, Intestinal protein OCI-5, MXR7, OCI5, OCI-5, secreted glypican-3, SGB, SGBS, SGBS1SDYS
Gene Symbol GPC3
Inhibitor Properties This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP1-85226. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL.
Research Category Cancer, Cell Biology, Cytoskeleton Markers, Signal Transduction, Stem Cells
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein Glypican 3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.