missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glypican-3 Mouse, Clone: 1G12 + GPC3/863, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 2719-MSM3-P1
This item is not returnable.
View return policy
Specifications
| Glypican-3 | |
| Cytoplasmic | |
| 1G12 + GPC3/863 | |
| Unconjugated | |
| NP_004475 | |
| Mouse | |
| 67kDa | |
| Protein A/G Purified | |
| RUO | |
| 2719 | |
| IgG |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 1mM PBS with 0.05% BSA and 0.05% azide. | |
| GPC3 | |
| Recombinant fragment containing amino acids 511-580 of human glypican-3 (1G12); Recombinant full-length human GPC3 protein (GPC3/863) | |
| 293T cells or Hepatocellular carcinoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction