missing translation for 'onlineSavingsMsg'
Learn More

Glypican 2 Antibody (300736) [DyLight 650], Novus Biologicals™

Product Code. 30736687 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736687 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736687 Supplier Novus Biologicals Supplier No. FAB2355W

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Glypican 2 Monoclonal antibody specifically detects Glypican 2 in Mouse samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glypican 2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 300736
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias 2410016G05Rik, GPC2
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Glypican 2, Lys27-Leu554, Accession # Q8BKV1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221914
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.