missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Glypican 1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547632
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse Glypican 1 is observed and less than 1% cross-reactivity with recombinant human (rh) Glypican 5 and rhGlypican 6 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation.
Specifications
| Glypican 1 | |
| Polyclonal | |
| Unconjugated | |
| GPC1 | |
| AI462976; glypican; glypican 1; glypican proteoglycan 1; glypican-1; Gpc1; GPI-anchored cell surface heparan sulfate proteoglycan; HSPG M12; HSPGM12; Secreted glypican-1 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 2817 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P35052 | |
| GPC1 | |
| Mouse myeloma cell line NS0-derived recombinant human Glypican 1 Asp24-Ser530. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction