missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Glypican 1 Antibody (Minomic patent anti-Glypican 1) - Humanized, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28590-50ug
This item is not returnable.
View return policy
Description
Glypican 1 Monoclonal antibody specifically detects Glypican 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Glypican 1 | |
| Monoclonal | |
| Unconjugated | |
| P35052 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 2817 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| Minomic patent anti-Glypican 1 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| FLJ38078, glypican, glypican 1, glypican proteoglycan 1, glypican-1 | |
| GPC1/ Glypican-1 | |
| 50 μg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction