missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glypican 1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-42718AF750
This item is not returnable.
View return policy
Description
Glypican 1 Polyclonal specifically detects Glypican 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Glypican 1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry | |
| FLJ38078, glypican, glypican 1, glypican proteoglycan 1, glypican-1 | |
| Glypican 1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of human glypican-1 protein. (Uniprot: P35052) | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 2817 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction