missing translation for 'onlineSavingsMsg'
Learn More

Glyphosate Stock Standard, SPEX CertiPrep™

Product Code. 15285796 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15285796 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15285796 Supplier SPEX Certiprep Supplier No. 547A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Glyphosate1071-83-62000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging 2mL amber prescored glass ampule
CAS 1071-83-6, 7732-18-5
CAS Min % 0.2, 99.8
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.2, 99.8
Methods For US EPA Methods 535, 547
Molecular Formula C3H8NO5P
MDL Number MFCD00055350
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key XDDAORKBJWWYJS-UHFFFAOYSA-N
SMILES OC(=O)CNCP(O)(O)=O
IUPAC Name 2-[(phosphonomethyl)amino]acetic acid
Solvent or Matrix H2O
Molecular Weight (g/mol) 169.07
PubChem CID 3496
Quantity 1 mL
ChEBI CHEBI:27744
Chemical Name or Material Glyphosate Stock Standard, USEPA Method 547
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.