missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glyphosate

Product Code. 16678263 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16678263 5 g 5g
17241202 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16678263 Supplier Thermo Scientific Chemicals Supplier No. 466180050
  • ONLINE EXCLUSIVE PRICE
    £151.00
    £135.65 / 5g
    Save £15.35
    10% Off
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1071-83-6 | C3H8NO5P | 169.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071-83-6
Molecular Formula C3H8NO5P
Molecular Weight (g/mol) 169.07
MDL Number MFCD00055350
InChI Key XDDAORKBJWWYJS-UHFFFAOYSA-N
IUPAC Name 2-[(phosphonomethyl)amino]acetic acid
SMILES OC(=O)CNCP(O)(O)=O

Specifications

Melting Point 230.0°C
Solubility Information Solubility aq. soln.: soluble. Other solubilities: insoluble in most organic solvents
Quantity 5 g
Formula Weight 169.07
Chemical Name or Material Glyphosate
compliance-icons compliance-icons
Product Identifier
  • Glyphosate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Glyphosate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.