missing translation for 'onlineSavingsMsg'
Learn More

Glyoxylic Acid Monohydrate, TRC

Product Code. 30134079
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134079 100 g 100g
30011613 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134079 Supplier Toronto Research Chemicals Supplier No. TRCG755000100G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-96-2
Molecular Formula C2H4O4
Molecular Weight (g/mol) 92.05
Synonym 2-Oxoacetic Acid Monohydrate, Glyoxalic Acid Monohydrate, Formylformic Acid Monohydrate, NSC 27785 Monohydrate, Oxalaldehydic Acid Monohydrate, Oxoacetic Acid Monohydrate, Oxoethanoic Acid Monohydrate, α-Ketoacetic Acid Monohydrate;
IUPAC Name 2,2-dihydroxyacetic acid
SMILES C(=O)C(=O)O.O

Specifications

Quantity 100 g
InChI Formula InChI=1S/C2H4O4/c3-1(4)2(5)6/h1,3-4H,(H,5,6)
Formula Weight 92.011
Chemical Name or Material Glyoxylic Acid Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.