missing translation for 'onlineSavingsMsg'
Learn More

Glyoxylic acid monohydrate, 98%, pure

Product Code. 10182480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10182480 25 g Plastic bottle 25g
10647652 100 g Plastic bottle 100g
10500941 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10182480 Supplier Thermo Scientific Acros Supplier No. 120170250
  • ONLINE EXCLUSIVE PRICE
    £27.10
    £24.65 / 25g
    Save £2.45
    9% Off
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 563-96-2 | C2H2O3·H2O | 92.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-96-2
Molecular Formula C2H2O3·H2O
Molecular Weight (g/mol) 92.06
MDL Number MFCD00127974
InChI Key MOOYVEVEDVVKGD-UHFFFAOYSA-N
Synonym glyoxylic acid monohydrate, glyoxylic acid hydrate, 2-oxoacetic acid hydrate, oxaldehydic acid hydrate, glyoxalic acid monohydrate, glyoxalate, glyoxylate hydrate, glyoxalic acid hydrate, oxoacetate monohydrate, glyoxylic acid-hydrate, oxoacetic acid hydrate
PubChem CID 15620607
IUPAC Name oxaldehydic acid;hydrate
SMILES C(=O)C(=O)O.O

Specifications

Melting Point 48.0°C to 52.0°C
Color White to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula HCOCO2H·H2O
Quantity 25 g
Solubility Information Solubility in water: freely soluble.
Formula Weight 92.06
Percent Purity 98%
Grade Pure
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Glyoxylic acid monohydrate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Glyoxylic acid monohydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.