missing translation for 'onlineSavingsMsg'
Learn More

Glyoxylic Acid (50% aqueous solution), TRC

Product Code. 17953975
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17953975 1 g 1g
17915055 10 g 10g
17972745 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17953975 Supplier Toronto Research Chemicals Supplier No. TRCG7549951G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-12-4
Molecular Formula C2 H2 O3
Molecular Weight (g/mol) 74.04
Synonym Acetic acid, 2-oxo-, Acetic acid, oxo- (9CI), Glyoxylic acid (8CI), 2-Oxoacetic acid, Formylcarboxylic acid, Formylformic acid, Glyoxalic acid, Glyoxo High Pure 50, NSC 27785, Oxalaldehydic acid, Oxoacetic acid, Oxoethanoic acid, α-Ketoacetic acid, Glyoxylic Acid (50% aqueous solution)
IUPAC Name oxaldehydic acid
SMILES OC(=O)C=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C2H2O3/c3-1-2(4)5/h1H,(H,4,5)
Formula Weight 74.0004
Chemical Name or Material Glyoxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.