missing translation for 'onlineSavingsMsg'
Learn More

Glyoxalic acid(50% in water), MedChemExpress

Product Code. 16401999
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16401999 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401999 Supplier Medchem Express Supplier No. HY79494

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Glyoxalic acid (NSC 27785) is an organic compound that is both an aldehyde and a carboxylic acid.

Glyoxalic acid (NSC 27785) is an organic compound that is both an aldehyde and a carboxylic acid.

Specifications

Chemical Name or Material Glyoxalic acid(50% in water)
Physical Form Liquid
CAS 298-12-4
Color Yellow
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C2H2O3
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym NSC 27785 Formylformic acid Oxalaldehydic acid
Solubility Information DMSO : 100 mg/mL (1350.62 mM; Need ultrasonic)
SMILES O=C(O)C=O
Molecular Weight (g/mol) 74.04
Formula Weight 74.04
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.