missing translation for 'onlineSavingsMsg'
Learn More

Glyoxalbis(2-hydroxyanil), 97%

Product Code. 10331514 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10331514 50 g Glass bottle 50g
10386460 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10331514 Supplier Thermo Scientific Acros Supplier No. 120160500
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1149-16-2 | C14H12N2O2 | 240.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1149-16-2
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
MDL Number MFCD00002183
InChI Key OMJNMRJMNUREOJ-KOBPNRPCSA-N
Synonym glyoxal bis 2-hydroxyanil, glyoxalbis 2-hydroxyanil, glyoxal bis o-hydroxyanil, glyoxal bis 2-hydroxyaniline, glyoxylidenebis 2-hydroxyaniline, unii-e2aw5xx09u, gbha, 2,2'-ethanediylidenedinitrilo bisphenol, phenol, 2,2'-1,2-ethanediylidenedinitrilo bis, 2,2'-ethanediylidenedinitrilo diphenol
PubChem CID 5356911
SMILES OC1=CC=CC=C1N\C=C\N=C1C=CC=CC1=O

Specifications

Melting Point 201°C to 205°C
Color Beige
Loss on Drying 0.5% max. (105°C)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 50 g
Solubility Information (0.5% in methanol) Clear
IUPAC Name 6-[(E)-2-(2-hydroxyanilino)ethenyl]iminocyclohexa-2,4-dien-1-one
Formula Weight 240.26
Percent Purity 97%
Physical Form Fine Powder
Chemical Name or Material Glyoxalbis(2-hydroxyanil)
Show More Show Less
compliance-icons
Product Identifier
  • Glyoxalbis(2-hydroxyanil)
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.