missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyoxalase II/HAGH Antibody (002), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90061AF350
This item is not returnable.
View return policy
Description
Glyoxalase II/HAGH Monoclonal specifically detects Glyoxalase II/HAGH in Human samples. It is validated for ELISA.
Specifications
| Glyoxalase II/HAGH | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 2 | |
| ELISA | |
| EC 3.1.2.6, GLO2hydroxyacyl glutathione hydrolase, Glx II, GLXII, Glyoxalase II, HAGH1GLX2, hydroxyacylglutathione hydrolase, hydroxyacylglutathione hydrolase, mitochondrial, hydroxyacylglutathione hydroxylase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Glyoxalase II/HAGH (Q16775-2; Met1-Asp260). | |
| 0.1 mL | |
| Lipid and Metabolism, Vision | |
| 3029 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction