missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyoxalase I Mouse anti-Human, Clone: CPTC-GLO1-3, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07312-100ug
This item is not returnable.
View return policy
Description
Glyoxalase I Monoclonal specifically detects Glyoxalase I in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
| Glyoxalase I | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase | |
| Recombinant full-length human Glyoxalase I protein | |
| 100 μg | |
| Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 2739 | |
| Store at 4°C. | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin) | |
| CPTC-GLO1-3 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction