missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyoxalase I inhibitor free base, MedChemExpress
Description
Glyoxalase I inhibitor (free base) is a potent Glyoxalase I (GLO1) inhibitor, candidate for anticancer agents.
Specifications
Specifications
| Chemical Name or Material | Glyoxalase I inhibitor free base |
| CAS | 174568-92-4 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H29BrN4O8S |
| Quantity | 5 mg |
| Solubility Information | DMSO : 110 mg/mL (190.49 mM; Need ultrasonic) |
| SMILES | N[C@H](C(OCC)=O)CCC(N[C@@H](CSC(N(O)C1=CC=C(Br)C=C1)=O)C(NCC(OCC)=O)=O)=O |
| Molecular Weight (g/mol) | 577.45 |
| Formula Weight | 577.45 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?