missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyoxalase I Antibody (Glo1a), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-19015B
This item is not returnable.
View return policy
Description
Glyoxalase I Monoclonal specifically detects Glyoxalase I in Human, Mouse samples. It is validated for Western Blot, Simple Western, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Glyoxalase I | |
| Monoclonal | |
| Biotin | |
| Q04760 | |
| GLO1 | |
| Full length human GLO1 protein [Swiss-Prot #Q04760]. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Glo1a | |
| Western Blot, Simple Western, ELISA, Immunocytochemistry/Immunofluorescence | |
| Aldoketomutase, EC 4.4.1.5, GLOD1, Glx I, GLYI, glyoxalase domain containing 1, glyoxalase Ialdoketomutase, Ketone-aldehyde mutase, lactoyl glutathione lyase, lactoylglutathione lyase, methylglyoxalase, S-D-lactoylglutathione methylglyoxal lyase | |
| Mouse | |
| 21 kDa | |
| 0.1 mL | |
| Cancer, Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 2739 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction