missing translation for 'onlineSavingsMsg'
Learn More

Glyoxal dimethyl acetal, 60% aq. soln.

Product Code. 11315015
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
Unit Size:
100g
2kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11315015 100 g 100g
11325015 500 g 500g
11335015 2000 g 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11315015 Supplier Thermo Scientific Alfa Aesar Supplier No. L18490.22
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51673-84-8 | C4H8O3 | 104.105 g/mol

Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glyoxal dimethyl acetal is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuffs.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51673-84-8
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.105
MDL Number MFCD00134410
InChI Key OGFKTAMJLKHRAZ-UHFFFAOYSA-N
Synonym acetaldehyde, dimethoxy, dimethoxyacetaldehyde, dimethoxy acetaldehyde, glyoxal dimethylacetal, glyoxal dimethyl acetal, 60 wt. % in h2o, acetaldehyde, 2,2-dimethoxy, 2,2-dimethoxyethanal, glyoxaldimethylacetal, dimethoxyethanal
PubChem CID 162650
IUPAC Name 2,2-dimethoxyacetaldehyde
SMILES COC(C=O)OC

Specifications

Density 1.15
Boiling Point 100°C to 102°C
Flash Point 102°C (216°F)
Refractive Index 1.414
Quantity 100 g
Beilstein 1850744
Solubility Information Fully miscible in water.
Formula Weight 104.11
Concentration or Composition (by Analyte or Components) 60% aq. soln.
Chemical Name or Material Glyoxal dimethyl acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.