missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glymidine Sodium, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Glymidine Sodium
Specifications
Specifications
| Chemical Name or Material | Glymidine Sodium |
| CAS | 3459-20-9 |
| Analyte or Component Names | Glymidine Sodium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H14 N3 O4 S . Na |
| Certifications/Compliance | ISO 17025 |
| Synonym | Benzenesulfonamide, N-[5-(2-methoxyethoxy)-2-pyrimidinyl]-, sodium salt (1:1), Benzenesulfonamide, N-[5-(2-methoxyethoxy)-2-pyrimidinyl]-, sodium salt (8CI,9CI), Sodium, [N-[5-(2-methoxyethoxy)-2-pyrimidinyl]benzenesulfonamido]- (7CI), BS 717, Glycanol, Glycodiazin, Glycodiazine, Glycodiazine sodium, Glyconormal, Glymidine, Glymidine sodium, Glymidine sodium salt, Gondafon, Lycanol, N-[5-(2-Methoxyethoxy)-2-pyrimidinyl]benzenesulfonamide sodium, Redul, Redul IF, Redul sodium, SH 717, Sodium 2-benzenesulfonylamino-5-(2-methoxyethoxy)pyrimidine, Sodium benzenesulfonamide, N-[5-(2-methoxyethoxy)-2-pyrimidinyl]-, Sodium glycodiazine, Sodium glymidine |
| InChI Formula | InChI=1S/C13H14N3O4S.Na/c1-19-7-8-20-11-9-14-13(15-10-11)16-21(17,18)12-5-3-2-4-6-12;/h2-6,9-10H,7-8H2,1H3;/q-1;+1 |
| SMILES | [Na+].COCCOc1cnc([N-]S(=O)(=O)c2ccccc2)nc1 |
| IUPAC Name | sodium;benzenesulfonyl-[5-(2-methoxyethoxy)pyrimidin-2-yl]azanide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction