missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycyrrhizic acid monoammonium salt, 98+%, Thermo Scientific Chemicals
£30.80 - £143.00
Chemical Identifiers
CAS | 53956-04-0 |
---|---|
Molecular Formula | C42H65NO16 |
Molecular Weight (g/mol) | 839.97 |
MDL Number | MFCD00167400,MFCD00167400 |
InChI Key | ILRKKHJEINIICQ-OOFFSTKBSA-N |
PubChem CID | 131676200 |
IUPAC Name | (2S,3S,4S,5R,6R)-6-[(2S,3R,4S,5S,6S)-2-[[(4aR,6aR,6bS,8aS,11R,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-2,3,4a,5,6,7,8,9,10,12,12a,14a-dodecahydro-1H-picen-3-yl]oxy]-6-carboxy-4,5-dihydroxyoxan-3-yl]oxy-3,4,5-trihydroxyoxane-2-carb |
SMILES | N.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O)O[C@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10590584
|
Thermo Scientific Acros
213750100 |
10 g | Glass bottle |
£30.80
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10696982
|
Thermo Scientific Acros
213750250 |
25 g | Glass bottle |
£53.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10771101
|
Thermo Scientific Acros
213751000 |
100 g | Glass bottle |
£143.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
53956-04-0 | |
839.97 | |
ILRKKHJEINIICQ-OOFFSTKBSA-N | |
(2S,3S,4S,5R,6R)-6-[(2S,3R,4S,5S,6S)-2-[[(4aR,6aR,6bS,8aS,11R,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-2,3,4a,5,6,7,8,9,10,12,12a,14a-dodecahydro-1H-picen-3-yl]oxy]-6-carboxy-4,5-dihydroxyoxan-3-yl]oxy-3,4,5-trihydroxyoxane-2-carb |
C42H65NO16 | |
MFCD00167400,MFCD00167400 | |
131676200 | |
N.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O)O[C@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O)C(O)=O |
Specifications
53956-04-0 | |
Authentic | |
C42H65NO16 | |
10,4369 | |
+ 51.50 | |
ILRKKHJEINIICQ-OOFFSTKBSA-N | |
(2S,3S,4S,5R,6R)-6-[(2S,3R,4S,5S,6S)-2-[[(4aR,6aR,6bS,8aS,11R,12aR,14aR,14bS)-11-carboxy-4,4,6a,6b,8a,11,14b-heptamethyl-14-oxo-2,3,4a,5,6,7,8,9,10,12,12a,14a-dodecahydro-1H-picen-3-yl]oxy]-6-carboxy-4,5-dihydroxyoxan-3-yl]oxy-3,4,5-trihydroxyoxane-2-carb | |
131676200 | |
98+% | |
Crystals or Powder |
209.0°C | |
98+% | |
MFCD00167400,MFCD00167400 | |
+ 51.50 (20.00°C c=1,EtOH 50% v/v) | |
Solubility in water: slightly soluble. Other solubilities: soluble in glacial acetic acid, ethanol,, soluble in ammonia water | |
N.CC1(C)[C@H](CC[C@@]2(C)[C@H]1CC[C@]1(C)[C@@H]2C(=O)C=C2[C@@H]3C[C@](C)(CC[C@]3(C)CC[C@@]12C)C(O)=O)O[C@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O[C@@H]1O[C@@H]([C@@H](O)[C@H](O)[C@H]1O)C(O)=O)C(O)=O | |
839.97 | |
839.98 | |
10% max. 18-alpha-glycyrrhizic acid (HPLC) | |
Glycyrrhizic acid monoammonium salt, 98% |
Safety and Handling
HYGROSCOPIC
missing translation for 'einecsNumber' : 258-887-7
missing translation for 'rtecsNumber' : LZ6500000
missing translation for 'tsca' : TSCA
RUO – Research Use Only