missing translation for 'onlineSavingsMsg'
Learn More

Glycyrrhizic acid, MedChemExpress

Product Code. 16429026
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16429026

Brand: Medchem Express HYN0184

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Glycyrrhizic acid is a triterpenoid saponinl, acting as a direct HMGB1 antagonist, with anti-tumor, anti-diabetic activities.

Glycyrrhizic acid is a triterpenoid saponinl, acting as a direct HMGB1 antagonist, with anti-tumor, anti-diabetic activities.

Specifications

Chemical Name or Material Glycyrrhizic acid
CAS 1405-86-3
Color White
Purity Grade Notes Research
Molecular Formula C42H62O16
Quantity 100 mg
Synonym Glycyrrhizin
Solubility Information DMSO : 500 mg/mL (607.59 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@]12[C@@](C(C=C3[C@]2(CC[C@]4(C)[C@@]3([H])C[C@](C(O)=O)(C)CC4)C)=O)([H])[C@@]5([C@@](C(C)([C@@H](O[C@]6([H])[C@@H]([C@H]([C@H](O)[C@@H](C(O)=O)O6)O)O[C@]7([H])O[C@@H]([C@@H](O)[C@H](O)[C@H]7O)C(O)=O)CC5)C)([H])CC1)C
Molecular Weight (g/mol) 822.93
Formula Weight 822.93
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.