missing translation for 'onlineSavingsMsg'
Learn More

Glycyrrhetic Acid, TRC

Product Code. 30393283
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30393283 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393283 Supplier Toronto Research Chemicals Supplier No. TRCG73500025G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 18beta-Glycyrrhetinic acid
CAS 471-53-4
Molecular Formula C30 H46 O4
Quantity 25 g
Synonym Enoxolone, Olean-12-en-29-oic acid, 3-hydroxy-11-oxo-, (3β,20β)-, (20β)-3β-Hydroxy-11-oxo-olean-12-en-29-oic acid, Olean-12-en-30-oic acid, 3β-hydroxy-11-oxo- (8CI), Uralenic acid (7CI), 18β-Glycyrrhetic acid, 18β-Glycyrrhetinic acid, Arthrodont, Biosone, Enoxolone, GM 1658, Glycyrrhetic acid, Glycyrrhetin, Glycyrrhetinic acid, NSC 35347, PO 12, STX 352, Subglycyrrhelinic acid, α-Glycyrrhetinic acid
InChI Formula InChI=1S/C30H46O4/c1-25(2)21-8-11-30(7)23(28(21,5)10-9-22(25)32)20(31)16-18-19-17-27(4,24(33)34)13-12-26(19,3)14-15-29(18,30)6/h16,19,21-23,32H,8-15,17H2,1-7H3,(H,33,34)/t19-,21-,22-,23+,26+,27-,28-,29+,30+/m0/s1
SMILES CC1(C)[C@@H](O)CC[C@@]2(C)[C@H]1CC[C@]3(C)[C@@H]2C(=O)C=C4[C@@H]5C[C@](C)(CC[C@]5(C)CC[C@@]34C)C(=O)O
IUPAC Name (2S,4aS,6aR,6aS,6bR,8aR,10S,12aS,14bR)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-3,4,5,6,6a,7,8,8a,10,11,12,14b-dodecahydro-1H-picene-2-carboxylic acid
Molecular Weight (g/mol) 470.68
Formula Weight 470.3396
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.