missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycylglycyl-L-tyrosine TFA Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Glycylglycyl-L-tyrosine TFA Salt |
| Analyte or Component Names | Glycylglycyl-L-tyrosine TFA Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13H17N3O5.(C2HF3O2) |
| Synonym | N-(N-Glycylglycyl)-L-tyrosine, N-(N-Glycylglycyl)tyrosine, Glycylglycyltyrosine, NSC 334194 |
| InChI Formula | InChI=1S/C13H17N3O5.C2HF3O2/c14-6-11(18)15-7-12(19)16-10(13(20)21)5-8-1-3-9(17)4-2-8;3-2(4,5)1(6)7/h1-4,10,17H,5-7,14H2,(H,15,18)(H,16,19)(H,20,21);(H,6,7)/t10-;/m0./s1 |
| Solution Type | Neat |
| SMILES | OC1=CC=C(C=C1)C[C@@H](C(O)=O)NC(CNC(CN)=O)=O.FC(F)(C(O)=O)F |
| IUPAC Name | glycylglycyl-L-tyrosine--2,2,2-trifluoroacetic acid (1/1) |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?