missing translation for 'onlineSavingsMsg'
Learn More

Glycylglycyl-L-tyrosine TFA Salt, TRC

Product Code. 30393966
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393966 25 mg 25mg
30394882 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30393966 Supplier Toronto Research Chemicals Supplier No. TRCG71861025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycylglycyl-L-tyrosine TFA Salt
Analyte or Component Names Glycylglycyl-L-tyrosine TFA Salt
Shipping Condition Room Temperature
Molecular Formula C13H17N3O5.(C2HF3O2)
Synonym N-(N-Glycylglycyl)-L-tyrosine, N-(N-Glycylglycyl)tyrosine, Glycylglycyltyrosine, NSC 334194
InChI Formula InChI=1S/C13H17N3O5.C2HF3O2/c14-6-11(18)15-7-12(19)16-10(13(20)21)5-8-1-3-9(17)4-2-8;3-2(4,5)1(6)7/h1-4,10,17H,5-7,14H2,(H,15,18)(H,16,19)(H,20,21);(H,6,7)/t10-;/m0./s1
Solution Type Neat
SMILES OC1=CC=C(C=C1)C[C@@H](C(O)=O)NC(CNC(CN)=O)=O.FC(F)(C(O)=O)F
IUPAC Name glycylglycyl-L-tyrosine--2,2,2-trifluoroacetic acid (1/1)
Industry Type Pharmaceutical
Molecular Weight (g/mol) 409.32
Quantity 25 mg
Formula Weight 409.11
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.