missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycylglycine, 99+%

Product Code. 10379223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10379223 1 kg 1kg
10366460 25 g 25g
10540771 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10379223 Supplier Thermo Scientific Chemicals Supplier No. 120140010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 556-50-3 | C4H8N2O3 | 132.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-50-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.12
MDL Number MFCD00008130
InChI Key YMAWOPBAYDPSLA-UHFFFAOYSA-N
Synonym glycylglycine, n-glycylglycine, diglycine, gly-gly, glycine dipeptide, glycine, glycyl, glycyl-glycine, glycine, n-glycyl, gly2, 2-2-aminoacetamido acetic acid
PubChem CID 11163
ChEBI CHEBI:17201
IUPAC Name 2-[(2-aminoacetyl)amino]acetic acid
SMILES C(C(=O)NCC(=O)O)N

Specifications

Melting Point 262.0°C to 264.0°C
Absorbance (10% aq. soln.) (1 cm cell), (10% aq. soln.) (1 cm cell) at 0nm, 0.01 max. at 405nm
pH 5.5 to 6 (5% aq. soln.)
Color White
Chloride 100ppm max
Infrared Spectrum Authentic
Assay Percent Range 99+%
Loss on Drying 0.5% max. (105°C, 4 hrs)
Packaging Plastic Bottle
Linear Formula H2NCH2CONHCH2CO2H
Beilstein 04, 371
Merck Index 15, 4539
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 132g/L (20°C). Other solubilities: slightly soluble in ethanol, practically insoluble in ether
Quantity 1 kg
Formula Weight 132.12
Percent Purity ≥99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Glycylglycine, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.