missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Glycylglycine, 99%
288.90 GBP valid until 2024-03-30
Use promo code "21648" to get your promotional price.
214.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21648" to get your promotional price.
214.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Glycylglycine is as a buffering reagent and also used in biochemical research. It is the simplest of all dipeptides and is used as a starting template for preparation of more complex peptides. It is also employed as a practical buffer with a buffering range of 7.5 - 8.9 (at 25°C). Valuable agent for experiments investigating peptide transport and absorption.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Glycylglycine is as a buffering reagent and also used in biochemical research. It is the simplest of all dipeptides and is used as a starting template for preparation of more complex peptides. It is also employed as a practical buffer with a buffering range of 7.5 - 8.9 (at 25°C). Valuable agent for experiments investigating peptide transport and absorption.
Solubility
Soluble in water (132 g/L) at 20°C, and ethanol (slightly solube).
Notes
Store at room temperature. Incompatible with oxidizing agents.
Glycylglycine is as a buffering reagent and also used in biochemical research. It is the simplest of all dipeptides and is used as a starting template for preparation of more complex peptides. It is also employed as a practical buffer with a buffering range of 7.5 - 8.9 (at 25°C). Valuable agent for experiments investigating peptide transport and absorption.
Solubility
Soluble in water (132 g/L) at 20°C, and ethanol (slightly solube).
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
Melting Point | ∼220°C (decomposition) |
Beilstein | 1765223 |
Merck Index | 14,4503 |
Solubility Information | Soluble in water (132g/L) at 20°C,and ethanol (slightly solube). |
Formula Weight | 132.12 |
Percent Purity | 99% |
Quantity | 500 g |
Chemical Name or Material | Glycylglycine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'