missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Glycylglycine
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Molecular Formula: C4H8N2O3
Molecular Weight: 132.1
CAS Number: 556-50-3Specifications
Specifications
| CAS | 556-50-3 |
| Quantity | 1 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction