missing translation for 'onlineSavingsMsg'
Learn More

Glycyl-L-Glutamic Acid, Fisher BioReagents

Product Code. p-26160
Change view
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Amber Glass
Unit Size:
1g
500mg
Product Code. Quantity Packaging unitSize
10695195 500 mg Amber Glass 500mg
10587714 1 g Amber Glass 1g
2 options
This item is not returnable. View return policy

Product Code. 10695195

Brand: Fisher Bioreagents BP2522500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Glycyl-L-Glutamic Acid is used in biochemical research applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7412-78-4
Molecular Formula C7H12N2O5
Molecular Weight (g/mol) 204.182
InChI Key IEFJWDNGDZAYNZ-BYPYZUCNSA-N
Synonym glycyl-l-glutamic acid, n-glycylglutamic acid, gly-glu, n-glycyl-l-glutamic acid, s-2-2-aminoacetamido pentanedioic acid, 2s-2-2-aminoacetamido pentanedioic acid, l-glutamic acid, n-glycyl, glycyl-l-glutamicacid, gly-l-glu, gly-l-glu-oh
PubChem CID 99278
ChEBI CHEBI:73801
IUPAC Name (2S)-2-[(2-aminoacetyl)amino]pentanedioic acid
SMILES C(CC(=O)O)C(C(=O)O)NC(=O)CN

Specifications

Color White
Packaging Amber Glass
Quantity 500 mg
Formula Weight 204.18
Physical Form Fluffy Powder
Chemical Name or Material Glycyl-L-Glutamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.