missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLYCTK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103201
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GLCTK.
This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described.
Specifications
| GLYCTK | |
| Polyclonal | |
| Unconjugated | |
| GLYCTK | |
| 6230410P16Rik; C430006L12; glycerate kinase; GLYCTK; HBeAg binding protein 4; HBeAg-binding protein 2; HBeAg-binding protein 4; HBeAgBP4A; HBEBP2; HBEBP4; LP5910 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 132158, 235582, 684314 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q0VGK3, Q8IVS8, Q8QZY2 | |
| GLYCTK | |
| A synthesized peptide derived from human GLYCTK(Accession Q8IVS8), corresponding to amino acid residues H269-A319. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction