missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLYCTK Monoclonal Antibody (1B5)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA523022
This item is not returnable.
View return policy
Description
Peptide Sequence: VLSAAMQGDV KSMAQFYGLL AHVARTRLTP SMAGASVEED AQLHELAAEL QIPDLQLEEA LETMAWGRGP VCLLAGGEPT VQLQGSGRGG RNQELALRV.
This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described.
Specifications
| GLYCTK | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q8IVS8, Q8QZY2 | |
| GLYCTK | |
| GLYCTK (NP_660305.2, 332 a.a. ~ 430 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1B5 | |
| Unconjugated | |
| GLYCTK | |
| 6230410P16Rik; C430006L12; glycerate kinase; GLYCTK; HBeAg binding protein 4; HBeAg-binding protein 2; HBeAg-binding protein 4; HBeAgBP4A; HBEBP2; HBEBP4; LP5910 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 132158, 235582 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction