missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLYCTK Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32317
This item is not returnable.
View return policy
Description
GLYCTK Polyclonal specifically detects GLYCTK in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| GLYCTK | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| GLYCTK | |
| Recombinant protein encompassing a sequence within the center region of human GLYCTK. The exact sequence is proprietary. | |
| 0.1 mL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.54mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| EC 2.7.1.31, glycerate kinase, GLYCTK1, HBeAg-binding protein 2, HBeAg-binding protein 4, HBeAgBP4A, HBEBP2, HBEBP4HBeAg binding protein 4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 132158 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction