missing translation for 'onlineSavingsMsg'
Learn More

Glycosylated Primaquine, TRC

Product Code. 30400495
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400495 2.5 mg 2.5mg
30406983 25 mg 25mg
30407338 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400495 Supplier Toronto Research Chemicals Supplier No. TRCG4280152.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycosylated Primaquine
Analyte or Component Names Glycosylated Primaquine
Shipping Condition Room Temperature
Molecular Formula C21 H31 N3 O6
Synonym (2S,3R,4R,5S)-2-(Hydroxymethyl)-6-((4-((6-methoxyquinolin-8-yl)amino)pentyl)amino)tetrahydro-2H-pyran-3,4,5-triol
InChI Formula InChI=1S/C21H31N3O6/c1-12(24-15-10-14(29-2)9-13-6-4-7-22-17(13)15)5-3-8-23-21-20(28)19(27)18(26)16(11-25)30-21/h4,6-7,9-10,12,16,18-21,23-28H,3,5,8,11H2,1-2H3/t12?,16-,18-,19+,20-,21?/m0/s1
Solution Type Neat
SMILES COc1cc(NC(C)CCCNC2O[C@@H](CO)[C@H](O)[C@@H](O)[C@@H]2O)c3ncccc3c1
IUPAC Name (2S,3R,4R,5S)-2-(hydroxymethyl)-6-[4-[(6-methoxyquinolin-8-yl)amino]pentylamino]oxane-3,4,5-triol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 421.487
Quantity 2.5 mg
Formula Weight 421.221
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.