missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Glycopyrronium bromide

Product Code. 17231222 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17231222 1 g 1g
17241222 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17231222 Supplier Thermo Scientific Chemicals Supplier No. 466320010
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 596-51-0
Molecular Formula C19H28BrNO3

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: soluble. Other solubilities: insoluble in dmso
Melting Point 193.0°C to 198.0°C
Formula Weight 398.34
Chemical Name or Material Glycopyrronium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.