missing translation for 'onlineSavingsMsg'
Learn More

GP6, eFluor 660, clone: HY101, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-9813-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15510147

  • £134.57 / 100 tests


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The HY101 monoclonal antibody reacts with the glycoprotein VI (GPVI), a 62-kDa type-I transmembrane glycoprotein that is expressed on mature megakaryotes and platelets. GPVI associates with the Fc receptor (FcR) gamma-chain to form a high affinity receptor for collagen. GPVI has also been reported to bind convulxin (Cvx). Phosphorylation of the ITAMs present on the FcR gamma-chain leads to recruitment of Src family kinases and activation of downstream signal transduction. GPVI has been reported to play a critical role in platelet activation and aggregation.Applications Reported: This HY101 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HY101 antibody has been pre-titrated and tested by flow cytometric analysis of normal human platelets. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Glycoprotein VI
HY101
eFluor 660
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
GPVI, GP6
IgG1
RUO
Primary
Human
Videos