missing translation for 'onlineSavingsMsg'
Learn More

Glycoprotein V/CD42d Antibody [DyLight 350], Novus Biologicals™

Product Code. 30736549 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736549 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736549 Supplier Novus Biologicals Supplier No. AF6990D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Glycoprotein V/CD42d Polyclonal antibody specifically detects Glycoprotein V/CD42d in Mouse samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glycoprotein V/CD42d
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias platelet glycoprotein V, CD42d, glycoprotein V (platelet), GPV
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Glycoprotein V/CD42d, Gln17-Gly522, Accession # NP_032174
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2814
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.