missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycoprotein V/CD42d Antibody (508209), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4249CL1
This item is not returnable.
View return policy
Description
Glycoprotein V/CD42d Monoclonal antibody specifically detects Glycoprotein V/CD42d in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Glycoprotein V/CD42d | |
| Monoclonal | |
| CoraFluor 1 | |
| platelet glycoprotein V, CD42d, glycoprotein V (platelet), GPV | |
| Mouse myeloma cell line NS0-derived recombinant human Glycoprotein V/CD42d, Gln17-Gly523, Accession # P40197 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 508209 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2814 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction