missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycophorin C Antibody (BRIC100), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65575AF532
This item is not returnable.
View return policy
Description
Glycophorin C Monoclonal specifically detects Glycophorin C in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| Glycophorin C | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| GYPC | |
| Synthetic peptide corresponding to amino acids in the cytoplasmic domain of GPC | |
| Primary | |
| Recognizes a 17 amino acid residue within the cytoplasmic domain of glycophorin C (GPC), which has recently been designated CD236. | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| BRIC100 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
| CD236, CD236 antigen, CD236R, GE, GLPC, glycoconnectin, glycophorin C (Gerbich blood group), glycophorin-C, glycophorin-D, Glycoprotein beta, GPCMGC126191, GPD, GYPD, MGC117309, MGC126192, PAS-2, PAS-2′, Sialoglycoprotein D | |
| Mouse | |
| 0.1 mL | |
| 2995 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction