missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycophorin C Antibody (BRIC10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66522APC
This item is not returnable.
View return policy
Description
Glycophorin C Monoclonal antibody specifically detects Glycophorin C in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
| Glycophorin C | |
| Monoclonal | |
| APC | |
| P04921 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| BRIC10 | |
| PBS | |
| CD236, CD236 antigen, CD236R, GE, GLPC, glycoconnectin, glycophorin C (Gerbich blood group), glycophorin-C, glycophorin-D, Glycoprotein beta, GPCMGC126191, GPD, GYPD, MGC117309, MGC126192, PAS-2, PAS-2', Sialoglycoprotein D | |
| Human erythrocytes | |
| 0.1 mL | |
| Cancer, Signal Transduction | |
| 2995 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction