missing translation for 'onlineSavingsMsg'
Learn More

Glycophorin C Antibody, Biotin, R&D Systems™

Product Code. 18610072 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18610072 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18610072 Supplier Novus Biologicals Supplier No. NBP297085B
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

Glycophorin C Polyclonal specifically detects Glycophorin C in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Glycophorin C
Applications Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias CD236, CD236 antigen, CD236R, GE, GLPC, glycoconnectin, glycophorin C (Gerbich blood group), glycophorin-C, glycophorin-D, Glycoprotein beta, GPCMGC126191, GPD, GYPD, MGC117309, MGC126192, PAS-2, PAS-2', Sialoglycoprotein D
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Glycophorin C (Accession#: NP_002092.1; Met1-Met57)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2995
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.