missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycophorin A Antibody (JC159), Novus Biologicals™
Mouse Monoclonal Antibody
£236.00 - £428.00
Specifications
| Antigen | Glycophorin A |
|---|---|
| Clone | JC159 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml, SDS-Page |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18283250
|
Novus Biologicals
NBP2-53415-20UG |
20 μg |
£236.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18290101
|
Novus Biologicals
NBP2-53415-100UG |
100 μg |
£428.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycophorin A Monoclonal specifically detects Glycophorin A in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Glycophorin A | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), SDS-Page | |
| Unconjugated | |
| Mouse | |
| Cancer, Signal Transduction | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 2993 | |
| Formalin fixed membrane from a Hairy cell leukemia | |
| Primary | |
| Store at 4C. |
| JC159 | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml, SDS-Page | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD235a antigen, glycophorin A (includes MN blood group), glycophorin A (MNS blood group), glycophorin Erik, glycophorin MiI, glycophorin MiIII, glycophorin MiV, glycophorin MiX, glycophorin SAT, glycophorin Sta type C, glycophorin-A, GPA, GPErik, GpMiIII, GPSAT, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), Mi.V glycoprotein (24 AA), MN sialoglycoprotein, MNS, PAS-2, recombinant glycophorin A-B Miltenberger-DR | |
| GYPA | |
| IgG1 κ | |
| Protein A or G purified | |
| 39 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title